Description
- Friction rods and pads made from various materials.
- All the essential materials for student exploration in electrostatics
- Topics include static electricity, polarity, and friction studies
- Use a variety of friction rods, friction pads, and pith balls
- The combination of materials will determine the charge distribution after contact is made
Specifications
Specifications
| Product Type | Friction Rods and Pads |
| Description | Pad |
| Material | Silk |
| For Use With (Application) | Friction Demonstrations |
| Length (English) | 12 in. |