Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frizzled 10/CD350 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982710
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2710 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2710 Supplier Bioss Supplier No. BS13216R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FZD10/Frizzled-10 is a 581 amino acid protein belonging to the G-protein coupled receptor Fz/Smo family and containing a signal peptide, a cysteine-rich domain in the N-terminal, seven transmembrane domains and a C-terminal PDZ domain-binding motif. It is involved in transduction, intercellular transmission of polarity information during tissue morphogenesis in differentiated tissues and as a receptor for Wnt proteins. FZD10 is expressed highly in placenta, fetal kidney, fetal lung, brain cerebellum, cerebral cortex, medulla and spinal cord with very low levels in total brain, frontal lobe, temporal lobe, putamen, adult brain, heart, lung and skeletal muscle.

Specifications

Antigen Frizzled 10/CD350
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FZD10
Gene Accession No. Q9ULW2
Gene Alias BOS_16452; CD350; CD350 antigen; frizzled 10, seven transmembrane spanning receptor; frizzled class receptor 10; frizzled family receptor 10; frizzled homolog 10; frizzled homolog 10 (Drosophila); frizzled homolog 10, pseudogene 1; Frizzled-10; Fz10; Fz-10; FZD10; Fzd10-ps1; FzE7; hFz10
Gene Symbols FZD10
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Frizzled 10/CD350.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11211
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.