Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Frizzled-10 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$208.00 - $448.50
Specifications
Antigen | Frizzled-10 |
---|---|
Clone | 1023101 |
Dilution | Immunohistochemistry 5-25 ug/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB3458SP
![]() |
R&D Systems
MAB3458SP |
25 μg |
Each for $208.00
|
|
|||||
MAB3458100
![]() |
R&D Systems
MAB3458100 |
100 μg |
Each for $448.50
|
|
|||||
Description
Frizzled-10 Monoclonal antibody specifically detects Frizzled-10 in Human samples. It is validated for ImmunohistochemistrySpecifications
Frizzled-10 | |
Immunohistochemistry 5-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
CD350, CD350 antigen, frizzled (Drosophila) homolog 10, frizzled homolog 10 (Drosophila), frizzled-10, Frizzled10, Fz10, Fz-10, FZD10, FzE7, hFz10 | |
Mouse myeloma cell line, NS0-derived human Frizzled-10, Ile21-Gly161, Accession # Q9ULW2 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1023101 | |
Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
Q9ULW2 | |
11211 | |
IgG2b | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title