Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Frizzled-2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518884
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518884 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518884 Supplier Invitrogen™ Supplier No. PA518884
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.

This intronless gene is a member of the frizzled gene family. Members of this family encode seven-transmembrane domain proteins that are receptors for the wingless type MMTV integration site family of signaling proteins. This gene encodes a protein that is coupled to the beta-catenin canonical signaling pathway. Competition between the wingless-type MMTV integration site family, member 3A and wingless-type MMTV integration site family, member 5A gene products for binding of this protein is thought to regulate the beta-catenin-dependent and -independent pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Frizzled-2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Fzd2
Gene Accession No. Q14332
Gene Alias AL033370; AW456835; Dfrizzled2; Drosophila polarity gene (frizzled) homologue; Drosophila polarity gene homolog 2; frizzled 2, seven transmembrane spanning receptor; frizzled class receptor 2; frizzled family receptor 2; frizzled homolog 2; frizzled homolog 2 (Drosophila); frizzled-10; Frizzled-2; Fz10; fz-10; Fz2; Fz-2; Fzd10; Fzd2; fzE2; hFz2; Mfz10; Mfz10a; mFz2; rFz2
Gene Symbols Fzd2
Host Species Goat
Immunogen Synthetic peptide sequence (RYATLEHPFHCPR) corresponding to the internal amino acids of FZD2 (aa 191-204).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2535
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.