Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frizzled-2, Rabbit anti-Human, Polyclonal, Millipore Sigma™

Catalog No. ABS1491100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS1491100 100 μL
ABS149125UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABS1491100 Supplier MilliporeSigma Supplier No. ABS1491100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detects Frizzled-2 using this rabbit polyclonal Anti-Frizzled-2 Antibody, Cat. No. ABS1491, tested for use in Western Blotting.

Frizzled-2 (UniProt: Q14332; also known as Fz-2, hFz2, FzE2) is encoded by the FZD2 gene (Gene ID: 2535) in human. Frizzled-2 is a multi-pass membrane protein that is widely expressed in adult tissues. In fetus, higher levels are expressed in the brain, lung, and kidney and lower levels are observed in liver. Frizzled serves as a receptor for Wnt proteins. Most of frizzled receptors are coupled to the beta-catenin canonical signaling pathway, which leads to the activation of disheveled proteins, inhibition of GSK-3 kinase, nuclear accumulation of beta-catenin and activation of Wnt target genes. In the presence of Wnt the Frizzled receptor is activated, which along with low-density lipoprotein receptor-related proteins (LRP) 5 and 6 stabilizes beta-catenin. The formation of Wnt-Frizzled-LRP6 complex, together with the recruitment of the scaffolding protein Dishevelled (Dvl), results in LRP6 phosphorylation and activation and the recruitment of the Axin complex to the receptors. This leads to the inhibition of Axin-mediated beta-catenin phosphorylation, which stabilizes beta-catenin. Frizzled-2 is synthesized with a signal peptide (aa 1-23), which is subsequently cleaved off in the mature form. Its Fz domain is localized to amino acids 34-153 and this domain is involved in its binding with Wnt ligands. Frizzled-2 can be ubiquitinated by RING-type E3 ubiquitin transferase ZNRF3 leading to its proteasomal degradation. Abnormalities in Wnt signaling are reported to promote birth defects, degenerative diseases, and cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Frizzled-2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Fz-2;hFz2;FzE2
Gene Symbols FZD2
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 13 amino acids from the N-terminal half of human Frizzled-2.
Molecular Weight of Antigen ∽60 kDa observed; 63.55 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody specifically detects human Frizzled -2. It targets an epitope with in 13 amino acids from the extracellular domain.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.