Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Frizzled-3 Antibody (169310) [Janelia Fluor™ 549], Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB1001I
Description
Frizzled-3 Monoclonal antibody specifically detects Frizzled-3 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOFSpecifications
Frizzled-3 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Flow Cytometry, Immunohistochemistry, CyTOF | |
169310 | |
Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
frizzled (Drosophila) homolog 3, frizzled homolog 3 (Drosophila), frizzled-3, Fz-3, hFz3 | |
Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-3, Met1-Val157, Accession # Q61086 | |
0.1 mL | |
GPCR, Growth and Development, Neuronal Cell Markers, Wnt Signaling Pathway | |
7976 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction