Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frizzled 4/CD344 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982711
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2711 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2711 Supplier Bioss Supplier No. BS13217R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FZD4 is a member of the frizzled gene family. Members of this family encode seven-transmembrane domain proteins that are receptors for the Wingless type MMTV integration site family of signaling proteins. Most frizzled receptors are coupled to the beta-catenin canonical signaling pathway. This protein may play a role as a positive regulator of the Wingless type MMTV integration site signaling pathway. A transcript variant retaining intronic sequence and encoding a shorter isoform has been described, however, its expression is not supported by other experimental evidence. Frizzled 4 is widely expressed, particularly in heart, skeletal muscle, ovary, and fetal kidney. Transcripts have also been identified in liver, kidney, pancreas, spleen, and fetal lung, and in smaller amounts in placenta, adult lung, prostate, testis, colon, and fetal brain.

Specifications

Antigen Frizzled 4/CD344
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FZD4
Gene Accession No. Q9ULV1
Gene Alias BOS_24395; CD344; CD344 antigen; EVR1; FEVR; frizzled 4, seven transmembrane spanning receptor; frizzled class receptor 4; frizzled family receptor 4; frizzled homolog 4; frizzled homolog 4 (Drosophila); frizzled receptor 4; Frizzled-4; Fz4; Fz-4; Fzd4; FZD4S; FzE4; GPCR; hFz4; mFz4; MGC34390; rFz4; WNT receptor frizzled-4
Gene Symbols FZD4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Frizzled 4/CD344.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8322
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.