Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Frizzled-5 Antibody (2D12) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP237451
Description
Frizzled-5 Monoclonal specifically detects Frizzled-5 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
Frizzled-5 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
C2orf31, chromosome 2 open reading frame 31, DKFZP434E2135, frizzled (Drosophila) homolog 5, frizzled homolog 5 (Drosophila), frizzled-5, Fz-5, FzE5, HFZ5, MGC129692, seven-transmembrane receptor frizzled-5, Wnt receptor | |
Mouse | |
64.5 kDa | |
0.1 mL | |
GPCR, Growth and Development, Neuronal Cell Markers, Signal Transduction, Wnt Signaling Pathway | |
7855 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Flow Cytometry, ELISA, CyTOF | |
2D12 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
FZD5 | |
Purified recombinant fragment of human FZD5 (AA:151-217) expressed in E. Coli. | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction