Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Frizzled-8 Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB1002439
Description
Frizzled-8 Polyclonal specifically detects Frizzled-8 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
Frizzled-8 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
frizzled (Drosophila) homolog 8, frizzled 8, frizzled family receptor 8, frizzled homolog 8 (Drosophila), frizzled-8, FZ-8, hFz8 | |
Goat | |
73 kDa | |
0.1 mg | |
GPCR, Growth and Development, Neuronal Cell Markers, Signal Transduction, Wnt Signaling Pathway | |
8325 | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
Western Blot 1 - 3 μg/mL, Immunohistochemistry 3 μg/mL, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 3 μg/mL, Peptide ELISA Detection limit 1:32000 | |
Q9H461 | |
FZD8 | |
Peptide with sequence C-SYPKQMPLSQV corresponding to C-Terminus according to NP_114072.1. | |
Affinity Purified | |
RUO | |
Primary | |
Expected from sequence similarity: Rat, Dog, Cow. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction