Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Frizzled-9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP216533
Description
Frizzled-9 Polyclonal specifically detects Frizzled-9 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Frizzled-9 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
CD349, CD349 antigen, frizzled (Drosophila) homolog 9, frizzled homolog 9 (Drosophila), frizzled-9, fz-9, FZD3fzE6, FzE6, hFz9 | |
Rabbit | |
64 kDa | |
0.1 mL | |
GPCR, Growth and Development, Neuronal Cell Markers, Signal Transduction, Wnt Signaling Pathway | |
8326 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
0.77mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
O00144 | |
FZD9 | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human Frizzled 9. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction