Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FRK Monoclonal Antibody (4E1)
GREENER_CHOICE

Catalog No. PIMA515547
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515547 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515547 Supplier Invitrogen™ Supplier No. MA515547
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15547 targets FRK in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15547 immunogen is purified recombinant fragment of human FRK expressed in E. Coli. MA5-15547 detects FRK which has a predicted molecular weight of approximately 58kDa.

FRK (PTK5) is a nuclear protein and may function during G1 and S phase of the cell cycle, suppressing growth. The human FRK gene maps to chromosome 6q21-q22.3 and encodes a 505 amino acid protein. The protein belongs to the TYR family of protein kinases. This tyrosine kinase is a nuclear protein and may function during G1 and S phase of the cell cycle and suppress growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FRK
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4E1
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene FRK
Gene Accession No. P42685
Gene Alias B-cell src-homology tyrosine kinase; beta-cell Src-homology tyrosine kinase; BSK; BSK/IYK; C85044; EC 2.7.1.112; EC 2.7.10.2; FRK; fyn related Src family tyrosine kinase; fyn-related kinase; fyn-related Src family tyrosine kinase; GASK; Gastrointestinal-associated kinase; gastrointestinal-associated tyrosine kinase; GTK; intestine tyrosine kinase; Iyk; Nuclear tyrosine protein kinase RAK; protein-tyrosine kinase 5; PTK5; PTK5 protein tyrosine kinase 5; RAK; src related tyrosine kinase; tyrosine-protein kinase FRK
Gene Symbols FRK
Host Species Mouse
Immunogen Purified recombinant fragment of human FRK expressed in E. Coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2444
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.