Learn More
FRK Mouse anti-Human, Clone: OTI5F3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF500629

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
FRK (Fyn-related kinase), also known as Rak. It is an epithelial tissue-specific kinase. The human FRK gene maps to chromosome 6q21-q22.3 and encodes a 505 amino acid protein. The protein belongs to the TYR family of protein kinases. This tyrosine kinase is a nuclear protein and may function during G1 and S phase of the cell cycle and suppress growth.Specifications
FRK | |
Monoclonal | |
Unconjugated | |
FRK | |
GTK, PTK5, RAK | |
Mouse | |
Affinity Chromatography | |
RUO | |
2444 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI5F3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P42685 | |
FRK | |
Full length human recombit protein of human FRK produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.