Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FRK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528373
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528373 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528373 Supplier Invitrogen™ Supplier No. PA528373
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (94%), Rat (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

FRK (PTK5) is a nuclear protein and may function during G1 and S phase of the cell cycle, suppressing growth. The human FRK gene maps to chromosome 6q21-q22.3 and encodes a 505 amino acid protein. The protein belongs to the TYR family of protein kinases. This tyrosine kinase is a nuclear protein and may function during G1 and S phase of the cell cycle and suppress growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FRK
Applications Western Blot
Classification Polyclonal
Concentration 0.81 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene FRK
Gene Accession No. P42685
Gene Alias B-cell src-homology tyrosine kinase; beta-cell Src-homology tyrosine kinase; BSK; BSK/IYK; C85044; EC 2.7.1.112; EC 2.7.10.2; FRK; fyn related Src family tyrosine kinase; fyn-related kinase; fyn-related Src family tyrosine kinase; GASK; Gastrointestinal-associated kinase; gastrointestinal-associated tyrosine kinase; GTK; intestine tyrosine kinase; Iyk; Nuclear tyrosine protein kinase RAK; protein-tyrosine kinase 5; PTK5; PTK5 protein tyrosine kinase 5; RAK; src related tyrosine kinase; tyrosine-protein kinase FRK
Gene Symbols FRK
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human FRK. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2444
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.