Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FRMD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553575
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553575 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553575 Supplier Invitrogen™ Supplier No. PA553575
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VVEASSKIQR EPPEVHRANI TQSRSSHSLN KQLIINMEPL QPLLPSPSEQ EEELPLGEGV PLPKEENISA PLISSSPVKA AREYEDPPSE EEDKIKEEPL TISELVYNPS ASLLPTPVDD DEIDMLFDCP SRLELEREDT DSFEDLEADE Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 83%.

Putative tumor suppressor gene that may be implicated in the origin and progression of lung cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FRMD3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FRMD3
Gene Accession No. A2A2Y4
Gene Alias 4.1O; 41O; band 4.1-like protein 4; band 4.1-like protein 4O; EPB41L4O; EPB41LO; FERM domain containing 3; FERM domain-containing protein 3; FRMD3; Ovary type protein 4.1; P410; protein 4.1O
Gene Symbols FRMD3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FRMD3. Recombinant protein control fragment (Product #RP-90475).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 257019
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.