Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FRMD4B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552677
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552677 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552677 Supplier Invitrogen™ Supplier No. PA552677
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LRGWYQRASG QKDQGHSPQT SFDSDRGSQR CLGFAGLQVP CSPSSRASLY SSVSSTNASG NWRTQLTIGL SDYETPAHSS YTSCYGNVYN PLPSPSRQYT EISQLDGTDG N Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 88%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen FRMD4B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FRMD4B
Gene Accession No. Q9Y2L6
Gene Alias 6030440G05Rik; C87375; FERM domain containing 4B; FERM domain-containing protein 4B; Frmd4b; GOBLIN; Golgi-associated band 4.1-like protein; GRP1 binding protein GRSP1; GRP1-binding protein GRSP1; GRSP1; KIAA1013; mKIAA1013; Nitzin; R74720
Gene Symbols FRMD4B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FRMD4B. Recombinant protein control fragment (Product #RP-90636).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23150
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.