Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FRMD6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121253
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121253 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121253 Supplier Invitrogen™ Supplier No. PA5121253
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: SH-SY5Y, HeLa, HepG2, MCF7, Mouse kidney, OVCAR8, Rat lung, Rat spleen. The target is usually found in the following locations: Cell membrane, Cytoplasm, Cytoplasmic side, Peripheral membrane protein. Immunogen sequence: KEWKKEASKV RQYEVTWGID QFGPPMIIHF RVQYYVENGR LISDRAARYY YYWHLRKQVL HSQCVLREEA YFLLAAFALQ ADLGNFKRNK HYGKYFEPEA YF.

The protein encoded by this gene interacts with signaling pathways to coordinately regulate cell growth, cell proliferation, apoptosis, autophagy, and cell migration. This tumor suppressor also enhances retinoblastoma 1 gene expression in cancer cells. Alternative splicing results in multiple transcript variants encoding distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FRMD6
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.95 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Frmd6
Gene Accession No. Q8C0V9, Q8VII0, Q96NE9
Gene Alias 2610019M19Rik; 4.1 ezrin radixin moesin (FERM)-containing protein; 4930488L10Rik; AW212977; c14_5320; C14orf31; EX1; expanded homolog; FERM domain containing 6; FERM domain containing 6; FERM domain-containing protein 6; FERM domain-containing protein 163SCII; FERM domain-containing protein 6; FERM-domain-containing protein 163SCII; Frmd6; MGC17921; Willin
Gene Symbols Frmd6
Host Species Rabbit
Immunogen A synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 122786, 257646, 319710
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.