Learn More
FRMD6 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305589AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
The protein encoded by this gene interacts with signaling pathways to coordinately regulate cell growth, cell proliferation, apoptosis, autophagy, and cell migration. This tumor suppressor also enhances retinoblastoma 1 gene expression in cancer cells. Alternative splicing results in multiple transcript variants encoding distinct isoforms.Specifications
FRMD6 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q8C0V9, Q96NE9 | |
FRMD6 | |
Between 572 and 622 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
Unconjugated | |
FRMD6 | |
4.1 ezrin radixin moesin (FERM)-containing protein, c14_5320, C14orf31, chromosome 14 open reading frame 31, EX1, expanded homolog, MGC17921, willin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
122786, 319710 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.