Learn More
Invitrogen™ FRMD7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551476
Description
Immunogen sequence: QVPRWSPIRA EERTSPHSYV EPTAMKPAER SPRNIRMKSF QQDLQVLQEA IARTSGRSNI NVGLEEEDPN LEDAFVCNIQ EQTPKRSQSQ SDMKTIRFPF GSEFRPLGPC PALSHKADLF TDMFAEQELP AVLMDQSTAE RYVASESSDS Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 79%.
Plays a role in neurite development, may be through the activation of the GTPase RAC1. Plays a role in the control of eye movement and gaze stability.
Specifications
| FRMD7 | |
| Polyclonal | |
| Unconjugated | |
| FRMD7 | |
| EG665849; FERM domain containing 7; FERM domain-containing protein 7; FRMD7; Gm1533; NYS; NYS1; RGD1562057; XIPAN | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 90167 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6ZUT3 | |
| FRMD7 | |
| Recombinant protein corresponding to Human FRMD7. Recombinant protein control fragment (Product #RP-92014). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.