Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FRMD7 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS8239R

Description
Plays a role in neurite development, may be through the activation of the GTPase RAC1. Plays a role in the control of eye movement and gaze stability.Specifications
FRMD7 | |
Polyclonal | |
Unconjugated | |
FRMD7 | |
FERM domain containing 7; FERM domain-containing protein 7; FRMD7; NYS; NYS1; XIPAN | |
Rabbit | |
Protein A | |
RUO | |
90167 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q6ZUT3 | |
FRMD7 | |
KLH conjugated synthetic peptide derived from human FRMD7. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction