Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FRMD8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551756
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551756 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551756 Supplier Invitrogen™ Supplier No. PA551756
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SREKHVLLGL RFQELSWDHT SPEEEEPILW LEFDGDSEGT PVNKLLKIYS KQAELMSSLI EYCIELSQAA EPAGPQDSAT GSPSDPSSSL APVQRPKLRR QGSVVSSRIQ HLSTIDYVED Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 88%.

FRMD8 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FRMD8
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FRMD8
Gene Accession No. Q9BZ67
Gene Alias 1200004M23Rik; 2310035N23Rik; 4931429L16Rik; AU018809; Band4.1 inhibitor LRP interactor; Bili; FERM domain containing 8; FERM domain-containing protein 8; FKSG44; FKSG44 protein; FRMD8; iRhom tail-associated protein; iTAP; RGD1310323
Gene Symbols FRMD8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FRMD8. Recombinant protein control fragment (Product #a href=.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83786
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.