Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FRMD8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531973
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531973 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531973 Supplier Invitrogen™ Supplier No. PA531973
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A549, HeLa, HepG2, HCT116, NIH-3T3. Predicted reactivity: Mouse (87%), Rat (89%), Rhesus Monkey (97%), Bovine (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

FRMD8 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FRMD8
Applications Western Blot
Classification Polyclonal
Concentration 0.77 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene FRMD8
Gene Accession No. Q3UFK8, Q9BZ67
Gene Alias 1200004M23Rik; 2310035N23Rik; 4931429L16Rik; AU018809; Band4.1 inhibitor LRP interactor; Bili; FERM domain containing 8; FERM domain-containing protein 8; FKSG44; FKSG44 protein; FRMD8; iRhom tail-associated protein; iTAP; RGD1310323
Gene Symbols FRMD8
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human FRMD8. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 67457, 83786
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.