Learn More
Invitrogen™ FRMPD1 Polyclonal Antibody

Description
A suggested positive control is K562 cell lysate. PA5-20832 can be used with blocking peptide PEP-0946.
Specifications
Specifications
| Antigen | FRMPD1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | FRMPD1 |
| Gene Accession No. | Q5SYB0 |
| Gene Alias | BC031840; FERM and PDZ domain containing 1; FERM and PDZ domain-containing protein 1; FERM domain-containing protein 2; FRMD2; Frmpd1; Kiaa0967; mKIAA0967; RP11-263I4.1 |
| Gene Symbols | FRMPD1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.