Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FRMPD2 Polyclonal Antibody
FRMPD2 Polyclonal Antibody
Supplier: Thermo Scientific PA580645
Description
FRMPD2 Polyclonal Antibody for IHC (P)

Specifications
FRMPD2 | |
Polyclonal | |
Unconjugated | |
FRMPD2 | |
ENSMUSG00000071536; FERM and PDZ domain containing 2; FERM and PDZ domain-containing protein 2; FRMPD2; Gm1582; Gm626; PDZ domain containing 5C; PDZ domain-containing protein 4; PDZ domain-containing protein 5C; PDZD5C; PDZK4; PDZK5C; predicted gene 626; RGD1565652 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
143162 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.03% ProClin 300; pH 7 | |
Q68DX3 | |
FRMPD2 | |
A synthetic peptide corresponding to the C-terminus of the Human FRMPD2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction