Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FRMPD3 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281718JF646
This item is not returnable.
View return policy
Description
FRMPD3 Polyclonal antibody specifically detects FRMPD3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
FRMPD3 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Janelia Fluor 646 | |
FERM and PDZ domain containing 3, FERM and PDZ domain-containing protein 3 | |
FRMPD3 antibody was raised against an 18 amino acid synthetic peptide from near the center of human FRMPD3. The immunogen is located within amino acids 690 - 740 of FRMPD3. Amino Acid Squence: PGFRDNSSDEDDPKRRAV | |
0.1 mL | |
Neuroscience | |
84443 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction