Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FRMPD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551844
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551844 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551844 Supplier Invitrogen™ Supplier No. PA551844
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MMKKEALLLI PNVLKVFLEN GQIKSFTFDG RTTVKDVMLT LQDRLSLRFI EHFALVLEYA GPEQNHKFLL LQDKQPLAYV VQRTHYHGMK CLFRISFFPK DPVELLRRDP AAFEYLYIQV ESGL Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 94%.

The FERM and PDZ domain containing (FRMPD) protein family consists of four proteins that contain a FERM (Four-point-one, erzin, radixin, moesin) domain and at least one PDZ (PSD-95/Discs large/Zonula-occuldens-1) domain. Unlike other members of the FRMPD family, FRMPD3 has no known function. It is most similar to FRMPD4, which is highly expressed in multiple regions of the brain and enriched in the postsynaptic density (PSD) fractions, it is possible that FRMPD3 also interacts with other PDZ domain-containing proteins in neural cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FRMPD3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FRMPD3
Gene Accession No. Q5JV73
Gene Alias B930041G04; EG245643; FERM and PDZ domain containing 3; FERM and PDZ domain-containing protein 3; FRMPD3; KIAA1817; RGD1563753; RP5-1070B1.1; similar to novel protein
Gene Symbols FRMPD3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FRMPD3. Recombinant protein control fragment (Product #RP-92312).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84443
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.