Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Therapak™ Frozen Biological Substance Category B Shipping Systems

Catalog No. 22130051 Shop All Therapak Corporation Products
Change view
Click to view available options
Capacity:
Holds 18 tubes
Holds 6 tubes
Holds 72 tubes
Holds 81 vials
Includes:
2 in. cryogenic box, Aqui-Pak pouch
3 in. cryogenic box, Aqui-Pak pouch
4-Lg canisters, 12 Aqui-Pak pouch
Large bag, 3 Aqui-Pak pouch
Large canister, 3 Aqui-Pak pouch
Medium canister, Aqui-Pak pouch
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity Includes
22-130-051 Holds 81 vials 3 in. cryogenic box, Aqui-Pak pouch
22-130-030 Holds 18 tubes Large canister, 3 Aqui-Pak pouch
22-130-031 Holds 81 vials 2 in. cryogenic box, Aqui-Pak pouch
22-130-423 Holds 72 tubes 4-Lg canisters, 12 Aqui-Pak pouch
22-130-029 Holds 6 tubes Medium canister, Aqui-Pak pouch
22-130-113 Holds 18 tubes Large bag, 3 Aqui-Pak pouch
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 22-130-051 Supplier Therapak™ Supplier No. 37808
Add to Cart
Add to Cart

Certified for ground and air shipments of Biological Substance Category B specimens

 

  • Designed exclusively for shipping frozen specimens in compliance with DOT and IATA Packaging Instruction 650 and 904
  • Suitable for overnight shipments
  • All systems include transport box; Biological Substance Category B and orientation arrow labels; 95kPa bag or canister or cryogenic box†; absorbent material; list of contents card; cushioning material; and packing instructions

 

Specifications

Quantity 1 Ea.
Includes 3 in. cryogenic box, Aqui-Pak pouch
Type Shipping System
Content And Storage Three in. cryogenic box; Aqui-Pak absorbent, cushioning, biohazard, Biological Substance Category B and arrow labels, list contents card, transport box
Capacity Holds 81 vials
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.