Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FRRS1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569436
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569436 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569436 Supplier Invitrogen™ Supplier No. PA569436
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.

Important modulator of glutamate signaling pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FRRS1L
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FRRS1L
Gene Accession No. Q9P0K9
Gene Alias 6430704M03Rik; brain protein CG-6; C11H9orf4; C9orf4; CG6; CG-6; DOMON domain-containing protein FRRS1L; EIEE37; Epb4.1l4b; erythrocyte protein band 4.1-like 4b; ferric chelate reductase 1 like; ferric-chelate reductase 1 like; ferric-chelate reductase 1-like; ferric-chelate reductase 1-like protein; FRRS1L; LOW QUALITY PROTEIN: DOMON domain-containing protein FRRS1L
Gene Symbols FRRS1L
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human FRRS1L.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23732
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.