Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FRRS1L Polyclonal Antibody
FRRS1L Polyclonal Antibody
Supplier: Thermo Scientific PA569437
Description
FRRS1L Polyclonal Antibody for Western Blot

Specifications
FRRS1L | |
Polyclonal | |
Unconjugated | |
FRRS1L | |
6430704M03Rik; brain protein CG-6; C11H9orf4; C9orf4; CG6; CG-6; DOMON domain-containing protein FRRS1L; EIEE37; Epb4.1l4b; erythrocyte protein band 4.1-like 4b; ferric chelate reductase 1 like; ferric-chelate reductase 1 like; ferric-chelate reductase 1-like; ferric-chelate reductase 1-like protein; FRRS1L; LOW QUALITY PROTEIN: DOMON domain-containing protein FRRS1L | |
Rabbit | |
Affinity chromatography | |
RUO | |
23732 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9P0K9 | |
FRRS1L | |
synthetic peptide directed towards the N terminal region of human FRRS1L. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction