Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FRRS1L Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567145
Description
Immunogen sequence: RHDSSYGTFAG EFYDLRYLSE EGYPFPTAPP VDPFAKIKVD DCGKTKGCFR YGKPGCNAET C Highest antigen sequence identity to the following orthologs - mouse 95%, rat 97%.
Important modulator of glutamate signaling pathway.
Specifications
FRRS1L | |
Polyclonal | |
Unconjugated | |
FRRS1L | |
6430704M03Rik; brain protein CG-6; C11H9orf4; C9orf4; CG6; CG-6; DOMON domain-containing protein FRRS1L; EIEE37; Epb4.1l4b; erythrocyte protein band 4.1-like 4b; ferric chelate reductase 1 like; ferric-chelate reductase 1 like; ferric-chelate reductase 1-like; ferric-chelate reductase 1-like protein; FRRS1L; LOW QUALITY PROTEIN: DOMON domain-containing protein FRRS1L | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23732 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P0K9 | |
FRRS1L | |
Recombinant Human FRRS1L. Recombinant protein control fragment (Product #RP-107791). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction