Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FRS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115253
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115253 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115253 Supplier Invitrogen™ Supplier No. PA5115253
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total FRS2.

Adapter protein that links activated FGR and NGF receptors to downstream signaling pathways. Plays an important role in the activation of MAP kinases and in the phosphorylation of PIK3R1, the regulatory subunit of phosphatidylinositol 3-kinase, in response to ligand-mediated activation of FGFR1. Modulates signaling via SHC1 by competing for a common binding site on NTRK1. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FRS2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene FRS2
Gene Accession No. Q8C180, Q8WU20
Gene Alias 4732458E18; C330018A15Rik; FGFR signalling adaptor; FGFR substrate 2; FGFR-signaling adaptor SNT; fibroblast growth factor receptor substrate 2; FRS1A; FRS2; Frs2a; FRS2alpha; FRS2-alpha; SNT; SNT1; SNT-1; SNT2; suc1-associated neurotrophic factor target 1
Gene Symbols FRS2
Host Species Rabbit
Immunogen A synthesized peptide derived from human FRS2(Accession Q8WU20), corresponding to amino acid residues S410-L460.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10818, 327826
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.