Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FRS3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA538921
Description
FRS3 (fibroblast growth factor receptor substrate 3), also known as FRS2B (FRS2-beta), is a 492 amino acid lipid-anchor adapter protein that contains one IRS-type PTB domain. Colocalizing to neural tissues with Tuj1, FRS3 functions as a feedback inhibitor of EGFR family members by preventing heterodimer formation between EGFR and ErbB2, thereby acting as a potential tumor suppressor. FRS3 is phosphorylated upon stimulation by FGF-2 or NGF and, acting as an adapter protein, links c-Fgr and NGF receptors to downstream signaling pathways. Interfering with the phosphorylation and nuclear translocation of ERK-2, FRS3 down-regulates ERK-2 expression. FRS3 likely interacts directly with GRB2, SH-PTP2, Flg, and Trk A, and may be involved in MAP kinase activation.
Specifications
FRS3 | |
Polyclonal | |
Unconjugated | |
FRS3 | |
Fibroblast growth factor receptor substrate 3, FGFR substrate 3, FGFR-signaling adaptor SNT2, Suc1-associated neurotrophic factor target 2, SNT-2, FRS3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10817 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O43559 | |
FRS3 | |
A synthetic peptide derived from the internal region of human FRS3 | |
100 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction