Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fructosamine-3-kinase-related Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299154UV
Description
Fructosamine-3-kinase-related Polyclonal specifically detects Fructosamine-3-kinase-related in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Fructosamine-3-kinase-related | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 350 | |
EC 2.7.1.-, FLJ12171, FN3KL, FN3K-related protein, FN3K-RP, fructosamine 3 kinase related protein, Fructosamine-3-kinase-related protein, ketosamine-3-kinase | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human Fructosamine-3-kinase-related. | |
0.1 mL | |
Protein Kinase | |
79672 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction