Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fructose, Granular, FCC, 98-102%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
57-48-7
C6H12O6
180.16
BJHIKXHVCXFQLS-UYFOZJQFSA-N
(3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
OC[C@@H](O)[C@@H](O)[C@H](O)C(=O)CO
57-48-7
C6H12O6
180.16
BJHIKXHVCXFQLS-UYFOZJQFSA-N
(3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
OC[C@@H](O)[C@@H](O)[C@H](O)C(=O)CO

F1092, 57-48-7, C6H12O6

Spectrum™ Fructose, Granular, FCC is a white, odorless crystalline solid that is extremely sweet and is the most water soluble of all sugars. It is used commercially as a sweetener in food and beverages due to its high sweetness. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Poly Pail
Loss on Drying 0.005
Quantity 2.5 kg
Ignition Residue 0.005
CAS Max % 1
Percent Purity 98 to 102%