Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fructose, Granular, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18602863 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
45 kg
Packaging:
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-863 4 x 2.5 kg Poly Bottle
18-602-860 500 g Poly Bottle
18-602-862 2.5 kg Poly Bottle
18-602-861 6 x 500 g Poly Bottle
18-602-864 12 kg Poly Bottle
18-602-865 45 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-602-863 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. FR1062.5KGCS4
Add to Cart
Add to Cart

FR106, 57-48-7, C6H12O6

Spectrum™ Chemical Fructose, Granular, USP is a white, odorless crystalline solid that is extremely sweet and is the most water soluble of all sugars. It is used as a sweetener in pharmaceutical formulations. Spectrum Chemical manufactured USP FRUCTOSE products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 57-48-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
InChI Key BJHIKXHVCXFQLS-UYFOZJQFSA-N
IUPAC Name (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
SMILES OC[C@@H](O)[C@@H](O)[C@H](O)C(=O)CO

Specifications

CAS Max % 100%
Loss on Drying 0.5%
Packaging Poly Bottle
Quantity 4 x 2.5 kg
Percent Purity 98 to 102%
Grade USP
Physical Form Crystals or Flakes
Chemical Name or Material Fructose, Granular
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.