Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fructosyl amino acid oxidase Antibody, Alexa Fluor™ 750, Novus Biologicals™

Sheep Polyclonal Antibody
Supplier: Novus Biologicals NBP177782AF750
Description
Fructosyl amino acid oxidase Polyclonal antibody specifically detects Fructosyl amino acid oxidase in Bacteria samples. It is validated for Western Blot, ELISASpecifications
Fructosyl amino acid oxidase | |
Polyclonal | |
Western Blot, ELISA | |
EC=1.5.3.-, FAOX-C | |
Fructosyl amino acid oxidase [from Corynebacterium sp. Expressed in E.coli] (Uniprot: Q8RIU8) | |
0.1 mL | |
Primary | |
Store at 4C in the dark. |
Western Blot, ELISA | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Sheep | |
Multi-step | |
RUO | |
Bacteria | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction