Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fructosyl amino acid oxidase Antibody, Biotin, Novus Biologicals™

Sheep Polyclonal Antibody
Supplier: Novus Biologicals NBP1777160.1MG
Description
Fructosyl amino acid oxidase Polyclonal specifically detects Fructosyl amino acid oxidase in Bacteria samples. It is validated for Western Blot, ELISA, Immunoprecipitation.Specifications
Fructosyl amino acid oxidase | |
Polyclonal | |
Western Blot 1:500 - 1:5000, ELISA 1:5000 - 1:20000, Immunoprecipitation 1:100 | |
Q8RIU8 | |
Sheep | |
Delipidation and Defibrination | |
RUO | |
Bacteria | |
Purified |
Western Blot, ELISA, Immunoprecipitation | |
Biotin | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide | |
faoxC | |
Fructosyl-Amino Acid Oxidase [from Corynebacterium sp. Expressed in E.coli] | |
0.1 mg | |
Primary | |
Store at 4C. Do not freeze. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction