Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FRY Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559655
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559655 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559655 Supplier Invitrogen™ Supplier No. PA559655
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EDVLFGFTNF LLREVNDMHH TLLDSSLKLL LQLLTQWKLV IQTQGKVYEQ ANKIRNSELI ANGSSHRIQS ERGPHCSVLH AVE Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 87%.

The FRY gene encodes a protein known as furry homolog (FRY), a gene that is highly conserved across species and plays vital roles in cell division and differentiation. FRY is involved in the regulation of mitotic spindle orientation and cellular polarity, crucial for proper cell division and tissue organization. It is found within various tissues and has implications in neurodevelopment and cancer biology, where it influences cell structural properties and signaling pathways. Alterations in FRY expression or mutations can disrupt normal cell function, leading to aberrant cellular morphogenesis and developmental defects. In cancer studies, FRY is associated with tumor suppression, where loss of function contributes to uncontrolled cell proliferation. This gene interacts with other cellular components, such as the cytoskeleton and signaling proteins, orchestrating complex cellular processes essential for maintaining homeostasis and development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FRY
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FRY
Gene Accession No. Q5TBA9
Gene Alias 13CDNA73; 214K23.2; bA207N4.2; bA37E23.1; C13orf14; CG003; FRY; FRY microtubule binding protein; furry homolog; protein furry homolog; WUGSC:H_2G3A.1
Gene Symbols FRY
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FRY. Recombinant protein control fragment (Product #RP-97919).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10129
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.