Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FRYL Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501563106
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-3106 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-3106 Supplier Bethyl Laboratories, Inc Supplier No. A302643AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

FRYL gene ontology annotations related to this gene include cell morphogenesis.

Specifications

Antigen FRYL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene FRYL
Gene Accession No. O94915
Gene Alias AF4p12, ALL1-fused gene from chromosome 4p12 protein, DKFZp686E205, FRY like, FRY-like, furry homolog-like, furry homolog-like (Drosophila), furry-like, KIAA0826, MOR2, mor2 cell polarity protein homolog, mor2 cell polarity protein homolog (S. pombe)
Gene Symbols FRYL
Host Species Rabbit
Immunogen Between 2600 and 2650
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285527, 72313
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.