Learn More
FRYL Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302644AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
FRYL gene ontology annotations related to this gene include cell morphogenesis.Specifications
FRYL | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
O94915 | |
FRYL | |
Between 2963 and 3013 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
FRYL | |
AF4p12, ALL1-fused gene from chromosome 4p12 protein, DKFZp686E205, FRY like, FRY-like, furry homolog-like, furry homolog-like (Drosophila), furry-like, KIAA0826, MOR2, mor2 cell polarity protein homolog, mor2 cell polarity protein homolog (S. pombe) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
285527 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.