Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FRZB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547793
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547793 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547793 Supplier Invitrogen™ Supplier No. PA547793
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.

The protein encoded by this gene is a secreted protein that is involved in the regulation of bone development. Defects in this gene are a cause of female-specific osteoarthritis (OA) susceptibility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FRZB
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene FRZB
Gene Accession No. P97401
Gene Alias cb291; Fiz; Fre; FREOS1; frezzled; Fritz; frizzled homolog-related; frizzled protein homolog; frizzled related protein; frizzled-related protein; frizzled-related protein 1; Frp; FRP-3; Frzb; Frzb precursor; FRZB1; FRZB-1; FRZB1sFRP-3; FRZB-PEN; fzb1; fzb-1; FZRB; hFIZ; id:ibd5075; ik:tdsubc_2e7; OS1; secreted frizzled-related protein 3; secreted frizzled-related sequence protein 3; SFRP3; sFRP-3; SFRP3frezzled; SRFP3; xx:tdsubc_2e7
Gene Symbols FRZB
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse sFRP-3 Ala33-Ser323.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20378
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.