Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FRZB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104120
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104120 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104120 Supplier Invitrogen™ Supplier No. PA5104120
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SFRP3.

The protein encoded by this gene is a secreted protein that is involved in the regulation of bone development. Defects in this gene are a cause of female-specific osteoarthritis (OA) susceptibility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FRZB
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FRZB
Gene Accession No. P97401, Q92765
Gene Alias cb291; Fiz; Fre; FREOS1; frezzled; Fritz; frizzled homolog-related; frizzled protein homolog; frizzled related protein; frizzled-related protein; frizzled-related protein 1; Frp; FRP-3; Frzb; Frzb precursor; FRZB1; FRZB-1; FRZB1sFRP-3; FRZB-PEN; fzb1; fzb-1; FZRB; hFIZ; id:ibd5075; ik:tdsubc_2e7; OS1; secreted frizzled-related protein 3; secreted frizzled-related sequence protein 3; SFRP3; sFRP-3; SFRP3frezzled; SRFP3; xx:tdsubc_2e7
Gene Symbols FRZB
Host Species Rabbit
Immunogen A synthesized peptide derived from human FRZB(Accession Q92765), corresponding to amino acid residues K285-N325.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20378, 2487, 295691
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.