Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FRZB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114374
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114374 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114374 Supplier Invitrogen™ Supplier No. PA5114374
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat brain tissue, rat kidney tissue, rat heart tissue, mouse brain tissue, mouse kidney tissue, mouse heart tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

The protein encoded by this gene is a secreted protein that is involved in the regulation of bone development. Defects in this gene are a cause of female-specific osteoarthritis (OA) susceptibility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FRZB
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene FRZB
Gene Accession No. P97401, Q92765
Gene Alias cb291; Fiz; Fre; FREOS1; frezzled; Fritz; frizzled homolog-related; frizzled protein homolog; frizzled related protein; frizzled-related protein; frizzled-related protein 1; Frp; FRP-3; Frzb; Frzb precursor; FRZB1; FRZB-1; FRZB1sFRP-3; FRZB-PEN; fzb1; fzb-1; FZRB; hFIZ; id:ibd5075; ik:tdsubc_2e7; OS1; secreted frizzled-related protein 3; secreted frizzled-related sequence protein 3; SFRP3; sFRP-3; SFRP3frezzled; SRFP3; xx:tdsubc_2e7
Gene Symbols FRZB
Host Species Rabbit
Immunogen E.coli-derived mouse FRZB recombinant protein (Position: N49-K311).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20378, 2487, 295691
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.