Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FRZB Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544188
Description
Peptide sequence: VVEVKEILKS SLVNIPRDTV NLYTSSGCLC PPLNVNEEYI IMGYEDEERS Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 92%.
The protein encoded by this gene is a secreted protein that is involved in the regulation of bone development. Defects in this gene are a cause of female-specific osteoarthritis (OA) susceptibility.
Specifications
FRZB | |
Polyclonal | |
Unconjugated | |
FRZB | |
cb291; Fiz; Fre; FREOS1; frezzled; Fritz; frizzled homolog-related; frizzled protein homolog; frizzled related protein; frizzled-related protein; frizzled-related protein 1; Frp; FRP-3; Frzb; Frzb precursor; FRZB1; FRZB-1; FRZB1sFRP-3; FRZB-PEN; fzb1; fzb-1; FZRB; hFIZ; id:ibd5075; ik:tdsubc_2e7; OS1; secreted frizzled-related protein 3; secreted frizzled-related sequence protein 3; SFRP3; sFRP-3; SFRP3frezzled; SRFP3; xx:tdsubc_2e7 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
2487 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q92765 | |
FRZB | |
Synthetic peptide directed towards the middle region of human FRZB. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction