Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FSBA Antibody (FSBA 2F5/5), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002656AF594
Description
FSBA Monoclonal specifically detects FSBA in Non-species specific samples. It is validated for ELISA, Immunohistochemistry.Specifications
FSBA | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
Mouse | |
Protein A purified | |
Epitope Tags | |
Non-species specific | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
FSBA 2F5/5 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
5′-(4-(fluorosulfonyl)benzoyl)adenosine, 5′-(4-fluorosulfonylbenzoyl)adenosine, 5′-FSBA, p-fluorosulfonylbenzoyl-5′-adenosine | |
Fluorylsulponylbenzoyladenosine (FSBA) | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction