Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FSBA Antibody (FSBA 2F5/5), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002656AF750
Description
FSBA Monoclonal specifically detects FSBA in Non-species specific samples. It is validated for ELISA, Immunohistochemistry.Specifications
| FSBA | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Mouse | |
| Protein A purified | |
| Epitope Tags | |
| Non-species specific | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| FSBA 2F5/5 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| 5′-(4-(fluorosulfonyl)benzoyl)adenosine, 5′-(4-fluorosulfonylbenzoyl)adenosine, 5′-FSBA, p-fluorosulfonylbenzoyl-5′-adenosine | |
| Fluorylsulponylbenzoyladenosine (FSBA) | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction