Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FSBP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555324
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555324 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555324 Supplier Invitrogen™ Supplier No. PA555324
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SNISEPTKKV MEMIPQISSF CLVRDRNHIQ SANLDEEAQA GTSSLQVMLD HHPVAITVEV KQEEDIKPPP PLVLNSQQSD TLEQREEHEL VHVMERSLSP Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 81%.

Fibrinogen silencer-binding protein, Function: Transcriptional repressor that down-regulates the expression of the fibrinogen gamma chain. Represses transcription of GSK3B gene promoter via its interaction with APBA1. Subunit structure: Interacts with APBA1 (via PDZ 1 and 2 domains). Subcellular location: Nucleus. Tissue specificity: Expressed in multiple tissues including brain. Miscellaneous: Intragenic, in the second intron of RAB54B gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FSBP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FSBP
Gene Accession No. O95073
Gene Alias fibrinogen silencer binding protein; fibrinogen silencer-binding protein; fibrinogen silencer-binding protein-like; FSBP; LOC102550523
Gene Symbols FSBP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FSBP. Recombinant protein control fragment (Product #RP-93950).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100861412
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.