Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FSCB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112968
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112968 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112968 Supplier Invitrogen™ Supplier No. PA5112968
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FSCB Polyclonal Antibody for Western Blot, IHC (P), ELISA

May be involved in the later stages of fibrous sheath biogenesis and spermatozoa capacitation. Inhibits ROPN1 and ROPN1L SUMOylation. Binds calcium. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FSCB
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene FSCB
Gene Accession No. A1EGX6, Q5H9T9
Gene Alias C14orf155; EG623046; fibrous sheath CABYR binding protein; fibrous sheath CABYR-binding protein; FSCB; PKA phosphorylated calcium and CABYR-binding protein; PKA-phosphorylated calcium and CABYR-binding protein
Gene Symbols FSCB
Host Species Rabbit
Immunogen Recombinant Human Fibrous sheath CABYR-binding protein (1-200AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 623046, 84075
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.