Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FSCN1 Monoclonal Antibody (OTI3B4), TrueMAB™, OriGene
SDP

Catalog No. 501681477
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-1477 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-1477 Supplier OriGene Supplier No. TA807293
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Human fascin is a highly conserved actin-bundling protein. Fascin, encoded by the human homolog for sn (hsn) gene, has been localized to microspikes and stress fibers of cultured cells where it is thought to be involved in the formation of microfilament bundles. It is expressed predomitly in dendritic cells. Lymphoid cells, myeloid cells and plasma cells are negative. However. Reed Sternberg cells in Hodgkin′s lymphoma are positive for fascin staining. Epstein-Barr virus may induce expression of fascin in B cells.

Specifications

Antigen FSCN1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3B4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene FSCN1
Gene Accession No. Q16658
Gene Alias FAN1, HSN, SNL, p55
Gene Symbols FSCN1
Host Species Mouse
Immunogen Full length human recombit protein of human FSCN1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6624
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.