Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FSCN1 Monoclonal Antibody (OTI3D2), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA807295
Description
Human fascin is a highly conserved actin-bundling protein. Fascin, encoded by the human homolog for sn (hsn) gene, has been localized to microspikes and stress fibers of cultured cells where it is thought to be involved in the formation of microfilament bundles. It is expressed predomitly in dendritic cells. Lymphoid cells, myeloid cells and plasma cells are negative. However. Reed Sternberg cells in Hodgkin′s lymphoma are positive for fascin staining. Epstein-Barr virus may induce expression of fascin in B cells.Specifications
| FSCN1 | |
| Monoclonal | |
| Unconjugated | |
| FSCN1 | |
| FAN1, HSN, SNL, p55 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 6624, 683788 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3D2 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P85845, Q16658 | |
| FSCN1 | |
| Full length human recombit protein of human FSCN1 produced in HEK293T. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction