Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FSCN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142288
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142288 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142288 Supplier Invitrogen™ Supplier No. PA5142288
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: This antibody is expected to recognize both reported isoforms (NP_001070650.1 and NP_036550.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.

FSCN2 acts as an actin bundling protein. May play a pivotal role in photoreceptor cell-specific events, such as disk morphogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FSCN2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene FSCN2
Gene Accession No. Q32M02
Gene Alias A930022G03; Ahl8; C630046B20Rik; fascin 2; fascin 2, retinal; fascin actin-bundling protein 2, retinal; fascin homolog 2, actin-bundling protein, retinal; fascin homolog 2, actin-bundling protein, retinal (Strongylocentrotus purpuratus); fascin-2; Fscn2; Retinal fascin; RFSN; RP30
Gene Symbols FSCN2
Host Species Goat
Immunogen Peptide with sequence CHHRGSNQLDTNR.
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 238021
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.